Claisen Distillation Adapters, Chemglass
Supplier: Chemglass
CG-1015-14 CG-1015-01 CG-1015-19
80065-738EA 320.29 USD
80065-738 80065-734 80065-740
Claisen Distillation Adapters, Chemglass
Distillation Components Distillation Adapters
Adapters feature a thermometer neck on the vertical side tube for immersion thermometers
The inner drip tip joint on the sidearm and the top outer joint and lower inner joint on the vertical tube are all the same [ST] joint size.