Weller® Controlled-Output Irons, Cooper Tools, Apex
Supplier: Cooper Tools
W100P3 W60P3 W100PG
37698-668EA 536.41 USD
37698-668 37698-804 37698-666
Weller® Controlled-Output Irons, Cooper Tools, Apex
Soldering Supplies Soldering Irons
Portable line voltage irons with no base station
Additional screwdriver tips are available in different sizes and soldering temperatures. All are designed to provide maximum heat transfer from heater to tip. Tips feature a copper core electroplated with iron to extend the lifetime of the tip. Nickel and chromium behind the working surface protect the shank from corrosion and solder creep. All tips are individually bagged.
Use a closed loop method of controlling maximum tip temperature. Stainless steel modular plug-in heater for greater efficiency and versatility. Come with plug-in nichrome wound element, 1.8m (6') cord, iron stand, and 371°C (700°F) screwdriver tip. The 60W irons include tip 37691-248; the 100W irons include tip 37692-878. Electrical: 120V.
Certificaciones: UL and C-UL listed.