CP-722 Heavy-Duty Bleach Solution, CANI
Supplier: Cani
CAN-722-15 CAN-722-30 CAN-722.1 CAN-722-5 CAN-722-55
93001-334EA 485.3 USD
93001-334 93001-338 93001-332 93001-340 93001-342
CP-722 Heavy-Duty Bleach Solution, CANI
Cleaning Agents Detergents and Surface Cleansers
Phosphate-free solution formulated for the pharmaceutical, biotechnology, cosmetic, medical device, and food and beverage industries
Contains highly alkaline chlorine (as hypochlorite) for effectively removing a wide range of process and product residues including proteins, synthetic organics, and dyes. Safe for use on glass and stainless steel. Solution does not foam over a wide temperature range and will not cavitate recirculation pumps. Free-rinsing formulation provides clean, residue-free surfaces.