Surface-Cleanse/930® Concentrated Neutral Cleaner, International Products
Supplier: International Products
S-2020 S-2090-055 S-2001-12
82013-122EA 1214.67 USD
82013-122 82013-124 82013-754
Surface-Cleanse/930® Concentrated Neutral Cleaner, International Products
Cleaning Agents Detergents and Surface Cleansers
This free-rinsing, nonionic cleaner has a neutral pH and low conductivity, making it ideal for cleaning items with surfaces or functions adversely affected by ionic surfactants
Contains no CFCs, ODCs, phosphates, borates, halogens, silicates, or phenols. Cleaner is NSF-registered as an A1, general cleaner in and around food-processing areas and equipment.
For use in manual washing operations, ultrasonic baths, and mild agitation systems. Cleaner mixes instantly with water and is effective at removing greases, oils, and other soils from many surfaces. Highly concentrated formula is effective for most cleaning applications at 0.5–2% dilution.