LF2100® Concentrated Liquid Low-Foam Cleaner, International Products
Supplier: International Products
L-2120 L-2190-055 L-2101-12
82010-764EA 1084.94 USD
82010-764 82010-766 82010-762
LF2100® Concentrated Liquid Low-Foam Cleaner, International Products
Cleaning Agents Detergents and Surface Cleansers
Powerful aqueous cleaning concentrate designed for both industrial and laboratory machine-based washing systems
Cleaner is easier to use than powders or gels; mixes instantly with water, and should be used at 1–2% dilution. Contains no CFCs, ODCs, phosphates, borates, halogens, silicates, or phenols. Meets USDA guidelines for incidental contact with food processing equipment. Specially blended for use in automated cleaning systems.
Removes a wide range of soils including greases, oils, waxes, particulates, oxides, biological materials, adhesives, mineral scales, tars, inks, clays,More Product Information