Viton® O-Rings, Chemglass
Supplier: Chemglass
CG-305-258 CG-305-267 CG-305-348 CG-305-349 CG-305-359 CG-305-5-101 CG-305-107 CG-305-006 CG-305-007 CG-305-008 CG-305-009 CG-305-010 CG-305-011 CG-305-012 CG-305-013 CG-305-014 CG-305-018 CG-305-019 CG-305-020 CG-305-022 CG-305-024 CG-305-027 CG-305-110 CG-305-111 CG-305-112 CG-305-113 CG-305-114 CG-305-115 CG-305-116 CG-305-117 CG-305-118 CG-305-120 CG-305-121 CG-305-122 CG-305-126 CG-305-128 CG-305-129 CG-305-132 CG-305-136 CG-305-137 CG-305-146 CG-305-201 CG-305-204 CG-305-205 CG-305-206 CG-305-208 CG-305-210 CG-305-212 CG-305-214 CG-305-215 CG-305-217 CG-305-218 CG-305-219 CG-305-220 CG-305-222 CG-305-223 CG-305-225 CG-305-226 CG-305-229 CG-305-232 CG-305-240 CG-305-242 CG-305-243 CG-305-317 CG-305-331 CG-305-338 CG-305-341 CG-305-104 CG-305-108 CG-305-109 CG-305-017
89029-271PK 121.57 USD
89029-271 89029-272 89029-284 89029-286 89029-288 89029-299 89029-695 89029-153 89029-155 89029-157 89029-159 89029-161 89029-165 89029-167 89029-169 89029-171 89029-173 89029-175 89029-177 89029-179 89029-181 89029-183 89029-185 89029-187 89029-189 89029-191 89029-193 89029-195 89029-197 89029-199 89029-201 89029-203 89029-205 89029-207 89029-209 89029-211 89029-213 89029-215 89029-217 89029-219 89029-221 89029-223 89029-225 89029-227 89029-229 89029-231 89029-233 89029-235 89029-237 89029-239 89029-241 89029-243 89029-245 89029-247 89029-249 89029-251 89029-253 89029-255 89029-257 89029-263 89029-265 89029-267 89029-269 89029-274 89029-276 89029-278 89029-282 89029-691 89029-697 89029-699 89041-342
Viton® O-Rings, Chemglass
Seals Sealing Rings
Brown, low compression, 75 durometer Viton® O-rings are suitable for acids, aliphatic hydrocarbons, oxidizing agents, and all petroleum products
Note: Viton® O-ring 89029-223 is not a standard size.
They are especially good for vacuum service and low gas permeability. Temperature range: –30 to 200°C (–22 to 392°F).