Mininert™ Syringe Valves for Vials, VICI®
Supplier: Vici Precision Sampling
614160 614158 614250 644850 614161 614163 614170
62403-002PK 403.71 USD
62403-002 62403-000 62403-010 62403-014 62403-004 62403-006 62403-008
Mininert™ Syringe Valves for Vials, VICI®
Closures Chromatography Vial Closures
Mininert™ fluoropolymer resin valves are leak-tight closures for screw cap or crimp-top vials
The screw cap Mininert™ valve threads onto the vial. The crimp-top style slides down into the neck of the vial. It features a threaded flange which can be turned to secure a tight fit. A rubber gasket above the fluoropolymer resin valve stem (isolated from the contents) provides a seal for the needle when the valve is open. It prevents leakage or exposure of the contents during sampling.
A Mininert™ septum installing tool is available separately. This lets you quickly remove and replace needleMore Product Information