2-(2,4-dichlorophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

87605-5G 87605-25G
77496-888EA 90.64 USD
77496-888 77505-844
2-(2,4-dichlorophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(2,4-dichlorophenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀Cl₂O₂
MW: 233.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771256
CAS Number: 84946-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR