5-Bromo-2-butoxy-4-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

82884-5G 82884-25G
77496-674EA 176.13 USD
77496-674 77505-686
5-Bromo-2-butoxy-4-methylpyridine ≥95%
5-Bromo-2-butoxy-4-methylpyridine

Precaución: For research use only

Formula: C₁₀H₁₄BrNO
MW: 244.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28127205
CAS Number: 1289008-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR