4-Bromo-2-fluoro-3-(methylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

81365-5G 81365-25G
77496-572EA 170.98 USD
77496-572 77505-608
4-Bromo-2-fluoro-3-(methylthio)benzonitrile ≥97%
4-Bromo-2-fluoro-3-(methylthio)benzonitrile

Precaución: For research use only

Formula: C₈H₅BrFNS
Storage Temperature: Ambient
MDL Number: MFCD34768779
CAS Number: 1507538-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR