(3-Bromophenyl)(isopropyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

78170-5G 78170-25G
77496-414EA 166.86 USD
77496-414 77505-498
(3-Bromophenyl)(isopropyl)sulfane ≥97%
(3-Bromophenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁BrS
Storage Temperature: Ambient
MDL Number: MFCD26142711
CAS Number: 70398-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR