1-(5-Hexylthiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

69117-5G 69117-25G
77495-926EA 150.38 USD
77495-926 77505-136
1-(5-Hexylthiophen-2-yl)ethanone ≥97%
1-(5-Hexylthiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29085268
CAS Number: 115466-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR