2,6-Dichloro-3-(1,3-dioxolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

68138-5G 68138-25G
77495-846EA 91.67 USD
77495-846 77505-072
2,6-Dichloro-3-(1,3-dioxolan-2-yl)pyridine ≥97%
2,6-Dichloro-3-(1,3-dioxolan-2-yl)pyridine

Precaución: For research use only

Formula: C₈H₇Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD28400800
CAS Number: 1485111-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR