1-Chloro-2,4,5-trifluorobenzene ≥95%

Supplier: AOBChem USA

Warning

65364-5G 65364-25G
77495-676EA 170.98 USD
77495-676 77504-964
1-Chloro-2,4,5-trifluorobenzene ≥95%
1-Chloro-2,4,5-trifluorobenzene

Precaución: For research use only

Formula: C₆H₂ClF₃
MW: 166.528 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194467
CAS Number: 2367-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR