4,6-Dibromo-2-methyl-3-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

62375-5G 62375-25G
77495-398EA 100.94 USD
77495-398 77504-754
4,6-Dibromo-2-methyl-3-(trifluoromethyl)aniline ≥97%
4,6-Dibromo-2-methyl-3-(trifluoromethyl)aniline

Precaución: For research use only

Formula: C₈H₆Br₂F₃N
MW: 332.9431 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760813
CAS Number: 2764731-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR