Ethyl 3-(3-bromo-4-methoxyphenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

58563-5G 58563-25G
77495-228EA 131.84 USD
77495-228 77504-642
Ethyl 3-(3-bromo-4-methoxyphenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(3-bromo-4-methoxyphenyl)-3-hydroxypropanoate

Precaución: For research use only

Formula: C₁₂H₁₅BrO₄
MW: 303.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17064765
CAS Number: 1457750-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR