2-(2-Chloro-6-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

55356-5G 55356-25G
77495-048EA 131.84 USD
77495-048 77504-510
2-(2-Chloro-6-fluorophenyl)-1,3-dioxolane ≥97%
2-(2-Chloro-6-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06208908
CAS Number: 154650-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR