4-Bromo-3-(tert-butyl)phenol ≥95%

Supplier: AOBChem USA

Warning

53782-5G 53782-25G
77494-974EA 152.44 USD
77494-974 77504-448
4-Bromo-3-(tert-butyl)phenol ≥95%
4-Bromo-3-(tert-butyl)phenol

Precaución: For research use only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD15527202
CAS Number: 103414-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR