1-(Benzyloxy)-3-bromo-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

52889-5G 52889-25G
77494-894EA 134.93 USD
77494-894 77504-384
1-(Benzyloxy)-3-bromo-5-methylbenzene ≥95%
1-(Benzyloxy)-3-bromo-5-methylbenzene

Precaución: For research use only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28973295
CAS Number: 876299-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR