3-Bromo-5-(diethoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

52255-5G 52255-25G
77494-846EA 90.64 USD
77494-846 77504-350
3-Bromo-5-(diethoxymethyl)pyridine ≥97%
3-Bromo-5-(diethoxymethyl)pyridine

Precaución: For research use only

Formula: C₁₀H₁₄BrNO₂
MW: 260.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404032
CAS Number: 2624417-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR