2-Bromo-5-iodo-3-methylthiophene ≥95%

Supplier: AOBChem USA

Warning

46862-5G 46862-25G
77494-508EA 131.84 USD
77494-508 77504-114
2-Bromo-5-iodo-3-methylthiophene ≥95%
2-Bromo-5-iodo-3-methylthiophene

Precaución: For research use only

Formula: C₅H₄BrIS
Storage Temperature: Ambient
MDL Number: MFCD26516700
CAS Number: 157925-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR