1-Chloro-3-(methoxymethoxy)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

36327-5G 36327-25G
77493-564EA 80.34 USD
77493-564 77503-328
1-Chloro-3-(methoxymethoxy)-2-methylbenzene ≥95%
1-Chloro-3-(methoxymethoxy)-2-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30695689
CAS Number: 2159430-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR