1,1,1,3,3,3-Hexafluoropropan-2-yl methanesulfonate ≥95%

Supplier: AOBChem USA

Warning

345325-5G 345325-25G
77492-990EA 114.33 USD
77492-990 77502-804
1,1,1,3,3,3-Hexafluoropropan-2-yl methanesulfonate ≥95%
1,1,1,3,3,3-Hexafluoropropan-2-yl methanesulfonate

Precaución: For research use only

Formula: C₄H₄F₆O₃S
MW: 246.1281 g/mol
Storage Temperature: Ambient
CAS Number: 25236-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR