2-Chloro-3-methylthio-6-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

29971-5G 29971-25G
77491-550EA 183.34 USD
77491-550 77501-526
2-Chloro-3-methylthio-6-(trifluoromethyl)pyridine ≥97%
2-Chloro-3-methylthio-6-(trifluoromethyl)pyridine

Precaución: For research use only

Formula: C₇H₅ClF₃NS
MW: 227.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28347336
CAS Number: 1809161-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR