1,3-Dioxolane, 2-(5-bromo-2,4-difluorophenyl)- ≥95%

Supplier: AOBChem USA

Warning

29291-5G 29291-25G
77491-350EA 162.74 USD
77491-350 77501-350
1,3-Dioxolane, 2-(5-bromo-2,4-difluorophenyl)- ≥95%
1,3-Dioxolane, 2-(5-bromo-2,4-difluorophenyl)-

Precaución: For research use only

Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977313
CAS Number: 2377575-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR