2-(3-Chloro-4-methylphenyl)quinazolin-4-ol ≥95%

Supplier: AOBChem USA

Warning

241496-5G 241496-25G
77490-268EA 142.14 USD
77490-268 77500-446
2-(3-Chloro-4-methylphenyl)quinazolin-4-ol ≥95%
2-(3-Chloro-4-methylphenyl)quinazolin-4-ol

Precaución: For research use only

Formula: C₁₅H₁₁ClN₂O
MW: 270.7136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870425
CAS Number: 2969311-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR