2-(Methylsulfonyl)pyridine ≥96%

Supplier: AOBChem USA

Warning

223254-5G 223254-25G
77490-036EA 95.79 USD
77490-036 77500-280
2-(Methylsulfonyl)pyridine ≥96%
2-(Methylsulfonyl)pyridine

Precaución: For research use only

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11212372
CAS Number: 17075-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR