2-Chloro-4-(trifluoromethyl)benzoic acid methyl ester ≥95%

Supplier: AOBChem USA

Warning

19797-5G 19797-25G
77489-554EA 142.14 USD
77489-554 77499-880
2-Chloro-4-(trifluoromethyl)benzoic acid methyl ester ≥95%
2-Chloro-4-(trifluoromethyl)benzoic acid methyl ester

Precaución: For research use only

Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD12923259
CAS Number: 192805-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR