1-(Ethylthio)-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

173167-5G 173167-25G
77488-810EA 176.13 USD
77488-810 77499-318
1-(Ethylthio)-4-methylbenzene ≥95%
1-(Ethylthio)-4-methylbenzene

Precaución: For research use only

Formula: C₉H₁₂S
MW: 152.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00093776
CAS Number: 622-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR