1-Methyl-4-(propylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

166713-5G 166713-25G
77488-488EA 130.81 USD
77488-488 77499-106
1-Methyl-4-(propylthio)benzene ≥95%
1-Methyl-4-(propylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00093778
CAS Number: 24599-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR