2-Phenylbutan-2-ol ≥97%

Supplier: AOBChem USA

Warning

164620-5G 164620-25G
77488-292EA 176.13 USD
77488-292 77498-964
2-Phenylbutan-2-ol ≥97%
2-Phenylbutan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00004475
CAS Number: 1565-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR