Phenyl(propyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164543-5G 164543-25G
77488-240EA 176.13 USD
77488-240 77498-926
Phenyl(propyl)sulfane ≥97%
Phenyl(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₂S
MW: 152.2565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039936
CAS Number: 874-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR