2,4,5-Trifluoro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

161151-5G 161151-25G
77488-160EA 70.04 USD
77488-160 77498-854
2,4,5-Trifluoro-N,N-dimethylbenzamide ≥97%
2,4,5-Trifluoro-N,N-dimethylbenzamide

Precaución: For research use only

Formula: C₉H₈F₃NO
MW: 203.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23378879
CAS Number: 871657-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR