1-Methoxymethoxy-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

14508-5G 14508-25G
77487-804EA 131.84 USD
77487-804 77498-522
1-Methoxymethoxy-4-(trifluoromethyl)benzene ≥95%
1-Methoxymethoxy-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₉H₉F₃O₂
MW: 206.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19688630
CAS Number: 174265-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR