1-(5-Bromothiophen-2-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

125583-5G 125583-25G
77487-318EA 114.33 USD
77487-318 77498-122
1-(5-Bromothiophen-2-yl)-2,2,2-trifluoroethanone ≥95%
1-(5-Bromothiophen-2-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₆H₂BrF₃OS
MW: 259.044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380810
CAS Number: 34773-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR