1-(2-Fluoro-5-methylphenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

118268-5G 118268-25G
77487-034EA 67.98 USD
77487-034 77497-880
1-(2-Fluoro-5-methylphenyl)-2-methylpropan-1-ol ≥97%
1-(2-Fluoro-5-methylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222657
CAS Number: 1343177-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR