2,4-Dichloro-3-(trimethylsilyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

82953-5G 82953-25G
77496-680EA 60.77 USD
77496-680 77505-690
2,4-Dichloro-3-(trimethylsilyl)pyridine ≥97%
2,4-Dichloro-3-(trimethylsilyl)pyridine

Precaución: For research use only

Formula: C₈H₁₁Cl₂NSi
MW: 220.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30177938
CAS Number: 134031-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR