2-(2-Fluoro-4-methylphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

81394-5G 81394-25G
77496-574EA 90.64 USD
77496-574 77505-610
2-(2-Fluoro-4-methylphenyl)-1,3-dioxolane ≥97%
2-(2-Fluoro-4-methylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06210009
CAS Number: 773087-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR