2-Fluoro-1-methoxy-3-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

80792-5G 80792-25G
77496-550EA 129.78 USD
77496-550 77505-594
2-Fluoro-1-methoxy-3-(methoxymethoxy)benzene ≥95%
2-Fluoro-1-methoxy-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₉H₁₁FO₃
MW: 186.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768484
CAS Number: 2901066-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR