2-(2,5-Difluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

77024-5G 77024-25G
77496-362EA 60.77 USD
77496-362 77505-454
2-(2,5-Difluorophenyl)-1,3-dioxolane ≥97%
2-(2,5-Difluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₈F₂O₂
MW: 186.1554 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209065
CAS Number: 773101-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR