5-(Methylthio)-2-(pyrrolidin-1-yl)pyridine ≥95%

Supplier: AOBChem USA

Warning

69021-5G 69021-25G
77495-914EA 162.74 USD
77495-914 77505-128
5-(Methylthio)-2-(pyrrolidin-1-yl)pyridine ≥95%
5-(Methylthio)-2-(pyrrolidin-1-yl)pyridine

Precaución: For research use only

Formula: C₁₀H₁₄N₂S
MW: 194.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763741
CAS Number: 2807471-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR