3,4-Dibromo-2,5-diiodothiophene ≥97%

Supplier: AOBChem USA

Warning

66463-5G 66463-25G
77495-736EA 119.48 USD
77495-736 77505-000
3,4-Dibromo-2,5-diiodothiophene ≥97%
3,4-Dibromo-2,5-diiodothiophene

Precaución: For research use only

Formula: C₄Br₂I₂S
MW: 493.725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762599
CAS Number: 67027-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR