5-Bromo-1,2-difluoro-3-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

64264-5G 64264-25G
77495-562EA 100.94 USD
77495-562 77504-872
5-Bromo-1,2-difluoro-3-(methoxymethoxy)benzene ≥95%
5-Bromo-1,2-difluoro-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇BrF₂O₂
MW: 253.0407 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761691
CAS Number: 1692680-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR