2-Bromo-4-fluoro-1-((4-methoxybenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

60697-5G 60697-25G
77495-324EA 100.94 USD
77495-324 77504-702
2-Bromo-4-fluoro-1-((4-methoxybenzyl)oxy)benzene ≥97%
2-Bromo-4-fluoro-1-((4-methoxybenzyl)oxy)benzene

Precaución: For research use only

Formula: C₁₄H₁₂BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD11127638
CAS Number: 1019441-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR