2-Bromo-4-cyclohexyl-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60378-5G 60378-25G
77495-306EA 100.94 USD
77495-306 77504-688
2-Bromo-4-cyclohexyl-1-methoxybenzene ≥97%
2-Bromo-4-cyclohexyl-1-methoxybenzene

Precaución: For research use only

Formula: C₁₃H₁₇BrO
MW: 269.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759717
CAS Number: 107274-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR