1-(2-Chloro-3-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

56817-5G 56817-25G
77495-138EA 142.14 USD
77495-138 77504-582
1-(2-Chloro-3-(trifluoromethyl)phenyl)ethanol ≥95%
1-(2-Chloro-3-(trifluoromethyl)phenyl)ethanol

Precaución: For research use only

Formula: C₉H₈ClF₃O
MW: 224.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31707321
CAS Number: 1488356-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR