1-(Methoxymethoxy)-2,4-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

53880-5G 53880-25G
77494-980EA 90.64 USD
77494-980 77504-452
1-(Methoxymethoxy)-2,4-dimethylbenzene ≥95%
1-(Methoxymethoxy)-2,4-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₄O₂
MW: 166.217 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21195664
CAS Number: 1497234-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR