1,2,4-Trifluoro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53748-5G 53748-25G
77494-970EA 100.94 USD
77494-970 77504-446
1,2,4-Trifluoro-3-isopropoxybenzene ≥97%
1,2,4-Trifluoro-3-isopropoxybenzene

Precaución: For research use only

Formula: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD23113677
CAS Number: 192446-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR