2-Chloro-3-ethoxy-4-fluoro-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52468-5G 52468-25G
77494-866EA 152.44 USD
77494-866 77504-366
2-Chloro-3-ethoxy-4-fluoro-1-methylbenzene ≥97%
2-Chloro-3-ethoxy-4-fluoro-1-methylbenzene

Precaución: For research use only

Formula: C₉H₁₀ClFO
MW: 188.627 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751506
CAS Number: 1807258-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR