1-Bromo-2-chloro-3-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

47382-5G 47382-25G
77494-528EA 91.67 USD
77494-528 77504-132
1-Bromo-2-chloro-3-(methoxymethoxy)benzene ≥97%
1-Bromo-2-chloro-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₈BrClO₂
MW: 251.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30214721
CAS Number: 1936101-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR